missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhenium plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13817AE

 View more versions of this product

Catalog No. AA13817AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Rhenium
Clear liquid
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
23947
Liquid
Colorless
7440-15-5,7697-37-2,7732-18-5
Re
Nitric acid: 5.0%; Rhenium: 0.1%; Water: 94.9%
[Re]
186.207
CHEBI:49882
23 hPa (17mm Hg) at 20°C
100mL

Chemical Identifiers

7440-15-5
186.207
WUAPFZMCVAUBPE-UHFFFAOYSA-N
CHEBI:49882
[Re]
Re
MFCD00011195
23947
rhenium
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents