missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium(IV) sulfide, 99%

Manufacturer:  Alfa Aesar™ 8948209

 View more versions of this product

Catalog No. AA8948209



Specifications

Specifications

Rhenium (IV) sulfide
ReS2
rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium
S=[Re]=S
250.327
250.33
99%
Odorless
12038-63-0
MFCD00016307
USWJSZNKYVUTIE-UHFFFAOYSA-N
bis(sulfanylidene)rhenium
82864
Powder
7.506g/mL
10g

Chemical Identifiers

12038-63-0
250.327
USWJSZNKYVUTIE-UHFFFAOYSA-N
82864
S=[Re]=S
ReS2
MFCD00016307
rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium
bis(sulfanylidene)rhenium
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents