Rhenium(III) chloride, Alfa Aesar™
Manufacturer:  Alfa Aesar 1323001
 View more versions of this product
Catalog No. AA1323001
Specifications
Specifications
Rhenium(III) chloride
Cl3Re
Dark red powder
LOIHSHVELSAXQN-UHFFFAOYSA-K
trichlororhenium
83581
Powder
Moisture sensitive
Red
13569-63-6
MFCD00011196
rhenium iii chloride, rhenium trichloride, rhenium chloride recl3, rhenium 3+ trichloride, recl3, acmc-20ajss, rhenium chloride recl3
Cl[Re](Cl)Cl
292.557
292.56
Odorless
Hydrate
0.25g
Chemical Identifiers
13569-63-6
292.557
LOIHSHVELSAXQN-UHFFFAOYSA-K
83581
Cl[Re](Cl)Cl
Cl3Re
MFCD00011196
rhenium iii chloride, rhenium trichloride, rhenium chloride recl3, rhenium 3+ trichloride, recl3, acmc-20ajss, rhenium chloride recl3
trichlororhenium
Safety and Handling
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 236-987-1
Documents