Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium foil, 1.0mm (0.04 in.) thick, 99.97% (metals basis)

Chemical Identifiers

Chemical Name or Material Rhenium foil
CAS 7440-15-5
Name Note 1.0mm (0.04 in.) thick
MDL Number MFCD00011195
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13984FF

  SDS
alfa aesar™
13984FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

TSCA TSCA
Chemical Name or Material Rhenium foil
CAS 7440-15-5
Assay Percent Notes (metals basis)
Odor Odorless
Quantity 25x25mm
Physical Form Foil
Assay 99.97%
Weight ≈13.15g/25x25mm
MDL Number MFCD00011195
Name Note 1.0mm (0.04 in.) thick