missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium foil, 0.5mm (0.02 in.) thick, annealed, 99.97% (metals basis)

Manufacturer:  Alfa Aesar™ 13983FF

 View more versions of this product

Catalog No. AA13983FF



Specifications

Specifications

Rhenium foil
25 x 25mm
7440-15-5
Re
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
Odorless
rhenium
23947
Foil
≈6.58g/25x25mm
0.5mm (0.02 in.) thick, Annealed
(metals basis)
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
[Re]
186.207
CHEBI:49882
99.97%

Chemical Identifiers

7440-15-5
186.207
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
CHEBI:49882
[Re]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents