Please login to your online account to display your discounted pricing

Rhenium AA Standard, 1mL = 1mg Re (1,000ppm Re)NH4ReO4 in H2O, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Rhenium AA Standard
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 13598-65-7
CAS Min % 0.14
CAS Max % 0.15
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ARE1KW100

 
ricca chemical company
RIARE1KW100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ARE1KW500

 
ricca chemical company
RIARE1KW500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Rhenium AA Standard
CAS 7732-18-5
Shelf Life 24 months
Concentration or Composition (by Analyte or Components) 1mL = 1mg Re (1, 000ppm Re)NH4ReO4 in H2O
CAS Min % 98.0
CAS Max % 100.0
CAS 13598-65-7
Packaging Poly natural
CAS Min % 0.14
CAS Max % 0.15
Safety and Handling

24 months