missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhenium, 1% on 2.5mm alumina spheres

Manufacturer:  Alfa Aesar™ 4702318

 View more versions of this product

Catalog No. AA4702318



Specifications

Specifications

Rhenium
7440-15-5
MFCD00011195
[Re]
186.207
CHEBI:49882
Spheres
1% on alumina spheres, 2.5mm alumina spheres
Re
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
23947
50g

Chemical Identifiers

7440-15-5
186.207
WUAPFZMCVAUBPE-UHFFFAOYSA-N
CHEBI:49882
[Re]
Re
MFCD00011195
23947
rhenium
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents