Please login to your online account to display your discounted pricing

Rhein, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Rhein
Name Note 90%
CAS 478-43-3
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C15H8O6
Formula Weight 284.22
MDL Number MFCD00009618
Synonym 4, 5-Dihydroxyanthraquinone-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC338992500

  SDS
thermo fisher scientific
338992500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥88%
Physical Form Crystalline Powder
Purity Grade Technical Grade
Formula Weight 284.22
Quantity 250mg
Color Yellow-orange to dark brown
RTECS Number CA9516000
EINECS Number 207-521-4
Merck Index 15, 8299
Synonym 4, 5-Dihydroxyanthraquinone-2-carboxylic acid
MDL Number MFCD00009618
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Name Note 90%
CAS 478-43-3
Assay Percent Range 90%
Molecular Formula C15H8O6
Chemical Name or Material Rhein
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.