Please login to your online account to display your discounted pricing

Thermo Scientific™ Reversible Microtube Racks with Lid

Store samples securely and rapidly identify them with Thermo Scientific™ Reversible Microtube Racks with Lid: featuring a low-profile design with an alphanumeric grid.

$708.69 - $708.69

Specifications

Description Reversible Microtube Rack
Tube Size 0.2mL tubes on one side, 0.5mL tubes on the other
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    

14-754-33

 
thermo scientific™
860011
Blue Case of 12 Each for $708.69
Description & Specifications

Specifications

Description Reversible Microtube Rack
Tube Size 0.2mL tubes on one side, 0.5mL tubes on the other

Store samples securely and rapidly identify them with  Thermo Scientific™ Reversible Microtube Racks with Lid: featuring a stable, low-profile design with an alphanumeric grid.

  • Hold 0.2mL tubes on one side, 0.5mL tubes on the other
  • Lid protects samples and allows stacking
  • 100% Recycled plastic
  • Several colors available