missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BUCHI FlashPure ID C18 Flash Cartridges

Effective for non-polar compounds

$95.06 - $1,566.74

Specifications

For Use With (Application) Effective for Non-Polar Compounds
Material Silica - C18
Max. Pressure 200 psig
Includes 1 Pc.
Type FlashPure
View More Specs

Includes: 1 Piece

Products 8
Catalog Number Mfr. No. Bed Weight (Metric) Price Quantity  
Catalog Number Mfr. No. Bed Weight (Metric) Price Quantity  
05-404-806
View Documents
BUCHI
145152103
12 g
Each for $149.22
 
05-404-807
View Documents
BUCHI
145152104
40 g
Each for $226.22
 
05-404-808
View Documents
BUCHI
145152105
80 g
Each for $362.20
 
05-404-809
View Documents
BUCHI
145152990
4 g
Each for $95.06
 
05-404-810
View Documents
BUCHI
145152991
120 g
Each for $475.39
 
05-404-836
View Documents
BUCHI
145164430
330 g
Each for $1,566.74
 
05-404-857
View Documents
BUCHI
145171532
24 g
Each for $186.52
 
05-404-858
View Documents
BUCHI
145171538
220 g
Each for $1,020.30
 
Description

Description

High performance

  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch

Lower purification costs

  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications

Specifications

Effective for Non-Polar Compounds
200 psig
FlashPure
Silica - C18
1 Pc.
SDS