BUCHI FlashPure ID C18 Flash Cartridges
Effective for non-polar compounds
Manufacturer:  BUCHI 145164430
 View more versions of this product
Catalog No. 05-404-836
Includes: 1 Piece
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications
Specifications
330 g
1 Pc.
100 psig
Effective for Non-Polar Compounds
Silica - C18
FlashPure
Documents