RETSCH ZM200 Ultra Centrifugal Mill: Distance Sieves
For temperature-sensitive materials
Supplier: RETSCH 036470258
Specifications
Distance Sieve | |
Stainless Steel | |
Trapezoid holes |
ZM200 ultra centrifugal mil | |
0.75 mm |