RETSCH Test Sieves - 14 Mesh or Greater
Stainless steel with wire gauze
Manufacturer:  RETSCH 60150000053
 View more versions of this product
Catalog No. 08-417-96
Specifications Description & Specifications
Specifications
2 in.
8 in.
ASTM E11
8 in.
270 Mesh
Stainless Steel
5.08cm
53μm
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
Stainless Steel
No. 270
Test Sieve