Please login to your online account to display your discounted pricing

RETSCH Test Sieves - 14 Mesh or Greater


Stainless steel with wire gauze

Manufacturer: retsch  60150001180

 View more versions of this product

Catalog No. 08-418-14

  • / Each
 

Description & Specifications

Specifications

Product Type Test Sieve
Mesh Material Stainless Steel
Tyler Standard 16 Mesh
Diameter (English) Outer 8 in.
For Use With (Application) Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
Material Stainless Steel
U.S. Standard No. 16
Certifications/Compliance ASTM E11
Depth (Metric) 5.08cm
Pore Size 1.18mm
Depth (English) 2 in.
Diameter (Metric) Outer 8 in.