RETSCH Test Sieves - 14 Mesh or Greater
Stainless steel with wire gauze
Supplier: RETSCH 60150000090
Specifications
Stainless Steel | |
8 in. | |
5.08cm | |
Test Sieve | |
8 in. | |
170 Mesh |
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc. | |
2 in. | |
90μm | |
ASTM E11 | |
Stainless Steel | |
No. 170 |