Please login to your online account to display your discounted pricing
RETSCH PM 100 Planterary Ball Mill Grinding Jars
Specially designed for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
Manufacturer: retsch  014620223
 View more versions of this product
Catalog No. 08-418-166
  • / Each
Description & Specifications
Specifications
Includes Nonslip seating, built-in anti-rotation device, conical base centering, O-ring, dustproof seal
Capacity (Metric) 250mL
For Use With (Application) Ideal for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
Capacity (English) 8.45 oz.
Product Type Grinding Jar
For Use With (Equipment) PM 100 and PM 200 planetary ball mill
Capacity 8.45 oz., 250mL
Material Stainless Steel