RETSCH PM 100 Planterary Ball Mill Grinding Jars
Specially designed for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
Manufacturer:  RETSCH 014620149
 View more versions of this product
Catalog No. 08-418-164
Specifications Description & Specifications
Specifications
1.69 oz.
1.69 oz., 50mL
Nonslip seating, built-in anti-rotation device, conical base centering, O-ring, dustproof seal
PM 100 and PM 200 planetary ball mill
50mL
Ideal for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
Stainless Steel
Grinding Jar