Please login to your online account to display your discounted pricing

RETSCH Mixer Mill Grinding Balls


Stainless steel grinding balls for use in mixer mills

Manufacturer: retsch  053680062

 View more versions of this product

Catalog No. 08-417-63

  • / Each
 

Description & Specifications

Specifications

Diameter (English) Outside 0.78 in.
For Use With (Application) Ideal for increasing the degree of mixing, grinding
Diameter (Metric) Outside 2cm
Composition 84.5% Fe, 13% Cr
For Use With (Equipment) MM 200, 301 mixer mill, S 100 centrifugal ball mill and PM 400 planetary ball mill
Material Stainless Steel
Product Type Grinding Ball