Please login to your online account to display your discounted pricing

RETSCH Mixer Mill Grinding Balls


Stainless steel grinding balls for use in mixer mills

Manufacturer: retsch  053680036

 View more versions of this product

Catalog No. 08-417-61

  • / Each
 

Description & Specifications

Specifications

Composition 84.5% Fe, 13% Cr
Diameter (English) Outside 0.35 in.
Diameter (Metric) Outside 0.9cm
For Use With (Application) Ideal for increasing the degree of mixing, grinding
Material Stainless Steel
For Use With (Equipment) MM 200 mixer mill
Product Type Grinding Ball