Please login to your online account to display your discounted pricing
RETSCH CryoMill Grinding Jar
Helps provide efficient milling
$1,298.75 - $1,298.75
Specifications
Cap Lid Type Screw Top
Product Type Grinding Jar
For Use With (Application) Ideal for cryogenic grinding
Material Stainless Steel
View More Specs
Products
Catalog Number Mfr. No. Capacity For Use With (Equipment) Price Quantity    
08-418-233
retsch
014620290
0.16 oz., 5mL Adapter of CryoMill cryogenic grinder Each for $1,298.75
Description & Specifications
Specifications
Cap Lid Type Screw Top
Product Type Grinding Jar
For Use With (Application) Ideal for cryogenic grinding
Material Stainless Steel
  • Stainless-steel grinding jar with screw-top design
  • Grinding jar is continually cooled with liquid nitrogen from the integrated cooling system before and during the grinding process