Please login to your online account to display your discounted pricing

Alfa Aesar™ Retinoic acid, 98%

$43.24 - $103.82

Chemical Identifiers

Chemical Name or Material Retinoic acid
CAS 302-79-4
Molecular Formula C20H28O2
Formula Weight 300.44
MDL Number MFCD00001551
Beilstein 2057223
Merck Index 14,8165
Synonym Vitamin A acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4454002

  SDS
alfa aesar™
4454002
0.5g Each for $103.82

AA4454077

  SDS
alfa aesar™
4454077
0.1g Each for $43.24
Description & Specifications

Specifications

Formula Weight 300.44
Chemical Name or Material Retinoic acid
TSCA TSCA
Physical Form Crystalline
Assay 98%
Molecular Formula C20H28O2
Sensitivity Light Sensitive
MDL Number MFCD00001551
CAS 302-79-4
Beilstein 2057223
Merck Index 14,8165
Synonym Vitamin A acid
Melting Point 180° to 182°C
Health Hazard 1 Warning
Quantity 0.5g
Health Hazard 2 GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VH6475000
Safety and Handling

Warning

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.