missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Retinoic acid, 98%

Manufacturer:  Alfa Aesar™ 4454077

 View more versions of this product

Catalog No. AA4454077



Specifications

Specifications

Retinoic acid
180°C to 182°C
302-79-4
MFCD00001551
14,8165
Soluble in ethanol,acetate,dimethyl formamide,dimethyl sulfoxide,chloroform,methanol,acetone and dichloromethane. Insoluble in water and ether.
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
300.442
CHEBI:15367
Crystalline
Light sensitive
0.1g
C20H28O2
2057223
airol
SHGAZHPCJJPHSC-YCNIQYBTSA-N
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
444795
300.44
98%

Chemical Identifiers

302-79-4
300.442
SHGAZHPCJJPHSC-YCNIQYBTSA-N
444795
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
C20H28O2
MFCD00001551
airol
CHEBI:15367
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.

P201-P202-P261-P264b-P270-P271-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335-H361

EINECSNumber : 206-129-0

RTECSNumber : VH6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents