Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7, Mouse Monoclonal Antibody.

$302.49 - $1,506.69

Specifications

Antigen Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7
Clone 51B7
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1883P

 
thermo scientific™ lab vision™
MS1883P
200μg/mL 1mL Each for $1,440.84

MS1883P0

 
thermo scientific™ lab vision™
MS1883P0
200μg/mL 100µL Each for $302.49

MS1883P1

 
thermo scientific™ lab vision™
MS1883P1
200μg/mL 500µL Each for $1,000.43

MS1883P1ABX

 
thermo scientific™ lab vision™
MS1883P1ABX
1mg/mL 100µL Each for $1,081.73

MS1883PABX

 
thermo scientific™ lab vision™
MS1883PABX
1mg/mL 200µL Each for $1,506.69
Description & Specifications

Specifications

Antigen Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7
Clone 51B7
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Isotype IgG1

Provide accurate, reproducible results with the Thermo Scientific™ Retinoblastoma (Rb) (Phospho-specific Serine) Ab-7, Mouse Monoclonal Antibody.

Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia. Phosphorylation of Rb at S608 depends on CDK4.

Host Species: Mouse

Clone: 51B7

Isotype: IgG1

Species Reactivity: Human and Mouse. Others not known.

Molecular Weight: 105kDa

Positive Control: Ls174T cells

Cellular Localization: Nuclear Ab-7 reacts with human and mouse protein phosphorylated on S608. It does not react with nonphosphorylate Rb or Rb phosphorylated at other sites.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO