missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Retinoblastoma (Rb) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Retinoblastoma (Rb) Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS107P1ABX

 View more versions of this product

Catalog No. MS107P1ABX


 

Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Retinoblastoma (Rb) Ab-1, Mouse Monoclonal Antibody.

Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.

Host Species: Mouse

Clone: 1F8; same as Rb1

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: aa 703-722

Immunogen: Recombinant human Rb protein

Molecular Weight: 105kDa

Positive Control: Ls174T, HT29, HUVEC cells; Normal colon, Breast CA and colon CA

Cellular Localization: Nuclear Ab-1 specifically stains the nuclei of BT-20 cells and primary human foreskin fibroblast (HFF) cells. It reacts with hyperphosphorylated and un (under) phosphorylated form of Rb protein. It shows no cross reaction with p107 and p130.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Retinoblastoma (Rb) Ab-1
Monoclonal
1mg/mL
Human
Ab without BSA/Azide (Frozen)
Recombinant human Rb protein
100μL
Cancer and Tumor Biology
Monoclonal
Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting
1F8; same as Rb1
Unlabeled
Concentrated
Murine
IgG1
RUO
Primary
Human