missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ RET Recombinant Human Protein DFS Item

The receptor tyrosine kinase RET is a member of the cadherin superfamily.

$2655.00 - $11190.00

Specifications

Accession Number NP_066124
For Use With (Application) Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Formulation 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Molecular Weight (g/mol) 79kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR7355A DFS Item
View Documents Promotion Details
Invitrogen™
PR7355A
100μg
Each for $2,655.00
 
PV4119 DFS Item
View Documents Promotion Details
Invitrogen™
PV4119
1mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

RET plays a crucial role in neural crest development, and can undergo oncogenic activation by cytogenetic rearrangement.

Specifications

Specifications

NP_066124
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
RET Kinase Family
Recombinant
Baculovirus
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
79kDa
RUO - research use only
RET
Human
GST Tag
Frozen
SDS
missing translation for 'documents'

missing translation for 'documents'