Invitrogen™ RET Recombinant Human Protein DFS Item
The receptor tyrosine kinase RET is a member of the cadherin superfamily.
Manufacturer:  Invitrogen™ LSPV4119
 View more versions of this product
Catalog No. PV4119
Description
Description
RET plays a crucial role in neural crest development, and can undergo oncogenic activation by cytogenetic rearrangement.
Specifications
Specifications
NP_066124
Frozen
79kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
RET Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
RET
Baculovirus
Human
GST Tag
Documents