RET [G691S] Recombinant Human Protein, Invitrogen™ DFS Item
Human RET G691S, C-terminal fragment, amino acids H658-S1114 (as in NCBI/Protein entry NP_066124.1) with a G691S mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
$614.00 - $9,380.00
Specifications
Host Species Human
Protein Tag GST Tag, His Tag
Recombinant Yes
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6214
View Documents
Life Technologies
LSPV6214
10μg Each for $614.00
PV6215
View Documents
Life Technologies
LSPV6215
100μg Each for $2,095.00
PV6216
View Documents
Life Technologies
LSPV6216
1mg Each for $9,380.00
Description
Description
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Human
GST Tag, His Tag
Yes