missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Resveratrol 99.0+%, TCI America™

Manufacturer:  TCI America R00715G

 View more versions of this product

Catalog No. R00715G



Specifications

Specifications

Resveratrol
267°C
501-36-0
MFCD00133799
LUKBXSAWLPMMSZ-OWOJBTEDSA-N
5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
445154
228.25
≥99.0% (GC)
Yellow
5g
C14H12O3
resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol
C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O
228.247
CHEBI:45713
Crystal-Powder at 20°C

Chemical Identifiers

501-36-0
228.247
LUKBXSAWLPMMSZ-OWOJBTEDSA-N
445154
5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
C14H12O3
MFCD00133799
resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol
CHEBI:45713
C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O
Safety and Handling

Safety and Handling

RTECSNumber : CA8987000

TSCA : No

Recommended Storage : Refrigerator

Documents