missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Restek™ Top Taper Inlet Liner for Bruker/Varian GCs DFS Item

For Bruker/Varian GCs equipped with 1078/1079 inlets.

$615.33 - $665.80

Specifications

Diameter (Metric) Inner 0.34 cm
Diameter (Metric) Outer 0.5 cm
Liner Material Borosilicate Glass
For Use With (Equipment) Bruker/Varian GCs Equipped with 1078/1079 Inlets
Geometry Top taper
View More Specs

Products 4
Catalog Number Mfr. No. Deactivation Packaging Quantity Packing Material Price Quantity  
Catalog Number Mfr. No. Deactivation Packaging Quantity Packing Material Price Quantity  
06711194 View Documents Restek™
RK20901209.5
Intermediate Polarity 5 Pk. CarboFrit N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06711196 View Documents Restek™
RK20901211.5
Base Deactivated 5 Pk. Base Deactivated with Wool Each for $615.33
 
06711195 View Documents Restek™
RK20901210.5
Base Deactivated 5 Pk. Base Deactivated N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06711199 View Documents Restek™
RK20901216.5
Siltek 5 Pk. CarboFrit Each for $665.80
 
Description

Description

  • Material: Borosilicate Glass
Specifications

Specifications

0.34 cm
Borosilicate Glass
Top taper
Top taper inlet liner
0.5 cm
Bruker/Varian GCs Equipped with 1078/1079 Inlets
5.4 cm