Restek™ Stabilwax™-DA Capillary Columns, Part 2: 50m Length DFS Item
Reduce adsorption and improve responses for many basic compounds
Specifications
Coating Fused Silica
Functionality/Interaction Type Acidic
Packing Material Stabilwax-DA
Technique Crossbond
For Use With Free (underivatized) Acids, some Inorganic Acids analysis
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Particle Size Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Particle Size Price Quantity  
06709359 View Documents Restek™
RK11027105
0.32mm 0.25μm 50m 0.25μm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06709357 View Documents Restek™
RK11026105
0.25mm 0.25μm 50m 0.25μm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06709375 View Documents Restek™
RK11042105
0.32mm 0.5μm 60m 0.5μm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06709379 View Documents Restek™
RK11043105
0.53mm 0.5μm 60m 0.5μm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Polar phase; Crossbond™ base-deactivated Carbowax™ polyethylene glycol–for amines and basic compounds)
  • Application-specific columns for underivatized amines and other basic compounds, including alkylamines, diamines, triamines, nitrogen-containing heterocyclics. No need for column priming
  • Temperature range: 40° to 220°C (104°F to 428°F)
  • Fused silica
Avoid rinsing these columns with water or alcohols.
Specifications
Specifications
Fused Silica
Stabilwax-DA
Free (underivatized) Acids, some Inorganic Acids analysis
Polar
G25, G35
Acidic
Crossbond
Stainless Steel
Carbowax Polyethylene Glycol