missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Restek™ SPI Liners for Varian, With Buffer
For Bruker/Varian GCs equipped with SPI inlets.
$679.42 - $679.42
Specifications
Length (Metric) | 54 mm |
---|---|
Product Type Specs | SPI Liner |
Geometry | SPI with Buffer |
Includes | Buffer |
Diameter (Metric) Inner | 2.4 mm |
Catalog Number | Mfr. No. | Deactivation | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Deactivation | Quantity | Price | Quantity | ||||
06718095
|
Restek™
RK20851 |
Intermediate Polarity (IP) | 5/Pk. |
Pack of 5 Each for $679.42
|
|
||||
Description
- Geometry: SPI with Buffer
- Material: Borosilicate Glass
- Deactivation: Intermediate Polarity (IP)
- Liner ID: 2.4mm
- Length: 54mm
- OD: 4.6mm
Specifications
54 mm | |
SPI with Buffer | |
2.4 mm | |
Bruker/Varian GCs Equipped with SPI Inlets |
SPI Liner | |
Buffer | |
4.6 mm | |
Borosilicate Glass |