missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Restek™ SPI Liners for Varian, With Buffer
For Bruker/Varian GCs equipped with SPI inlets.
Supplier: Restek™ RK20851
Description
- Geometry: SPI with Buffer
- Material: Borosilicate Glass
- Deactivation: Intermediate Polarity (IP)
- Liner ID: 2.4mm
- Length: 54mm
- OD: 4.6mm
Specifications
SPI with Buffer | |
Intermediate Polarity (IP) | |
5/Pk. | |
2.4 mm | |
Bruker/Varian GCs Equipped with SPI Inlets |
Buffer | |
SPI Liner | |
54 mm | |
4.6 mm | |
Borosilicate Glass |