Restek™ Specialty Resprep™ RDX Cartridge DFS Item
Extraction of explosive compounds (EPA Method 8330) from water samples
Manufacturer:  Restek™ RK26093
Catalog No. 06713154
Specifications
Specifications
Extraction of Explosive Compounds (EPA Method 8330) From Water Samples
RDX
SPE Cartridge
HDPE, Polyethylene Frits
6 mL
Documents