Restek™ Rtx™-WAX Capillary Columns: With Guard or Cage DFS Item
With excellent selectivity compared to other Carbowax™ columns
$1,488.96 - $1,488.96
Specifications
Coating Fused Silica
Packing Material Rtx™-Wax
Technique Crossbond™
For Use With Alkenols, Glycols, Aldehydes analysis
Material Stainless Steel
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Particle Size Includes Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Particle Size Includes Price Quantity  
06709755 View Documents Restek™
RK12426280
0.25mm 60m 0.25μm Star cage, 4 in. coil N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06709764 View Documents Restek™
RK124386850
0.25mm 30m 0.5μm Cage N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06709782 View Documents Restek™
RK124556850
0.53mm 30m 1μm Cage Each for $1,488.96
Description
Description
Include guard column or cage.
  • Polyethylene glycol phase (Crossbond™ polyethylene glycol) with extended operating temperatures for the analysis of compounds having a wide volatility range
  • High polarity
  • Provide thermal stability with guaranteed low bleed to 250°C (482°F); low (20°C) minimum usable temperature
  • Fast and efficient for BTEX analysis
  • Include individual test chromatogram
FAMEs, Essential oils, Polar solvents
Specifications
Specifications
Fused Silica
Crossbond™
Stainless Steel
Intermediate to high polar
G14, G15, G16, G20, G39
Rtx™-Wax
Alkenols, Glycols, Aldehydes analysis
350°C
Carbowax™ Polyethylene Glycol