Restek™ Rtx™-VMS Capillary Columns: With Cage DFS Item
Provide complete separation of all U.S. EPA Method 8260 compounds in less than 18 minutes
$1,367.56 - $1,367.56
Specifications
Coating Fused Silica
Packing Material Rtx™-V MS
Technique Crossbond™
For Use With Volatile organic pollutants analysis
Material Stainless Steel
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Particle Size Includes Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Particle Size Includes Price Quantity  
06710489 View Documents Restek™
RK199156850
0.25mm 30m 1.4μm Cage Each for $1,367.56
06710492 View Documents Restek™
RK199196850
0.32mm 30m 1.8μm Cage N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06713259 View Documents Restek™
RK49914042
0.18mm 20m 1μm Cage N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06713260 View Documents Restek™
RK499146850
0.18mm 20m 1μm Cage N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
With cage.
  • Special polymer formulation features low bleed, improved selectivity, and faster analysis time for separation of volatile organic compounds by GC/MS
  • Excellent thermal stability
  • Individual test chromatogram is provided with each column
Specifications
Specifications
Fused Silica
Crossbond™
Stainless Steel
Proprietary
Rtx™-V MS
Volatile organic pollutants analysis
350°C