Restek™ RTX™-5 Capillary Columns: Length 10m DFS Item
Fused silica
Specifications
Coating Fused Silica
Packing Material Rtx-5
Technique Crossbond
For Use With Drugs, solvent impurities, pesticides, Hydrocarbons, PCB congeners, essential oils, semivolatiles analysis
Material Stainless Steel
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Particle Size Includes Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Particle Size Includes Price Quantity  
06713218 View Documents Restek™
RK412016850
0.1mm 10m 0.1μm Cage N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06713217 View Documents Restek™
RK41201051
0.1mm 10m 0.1μm N/A N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06713221 View Documents Restek™
RK41203051
0.1mm 10m 0.4μm N/A N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06713222 View Documents Restek™
RK412036850
0.1mm 10m 0.4μm Cage N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Low polarity phase; Crossbond™ (diphenyl dimethyl polysiloxane)
  • General-purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g., Aroclor mixes), essential oils, semivolatiles
  • Temperature range: -60°C to 350°C (-76°F to 662°F)
  • Equivalent to USP G27 and G36 phases
Specifications
Specifications
Fused Silica
Crossbond
Stainless Steel
5% Diphenyl/95% Dimethyl Polysiloxane
Rtx-5
Drugs, solvent impurities, pesticides, Hydrocarbons, PCB congeners, essential oils, semivolatiles analysis
Low polar
G27, G36