Restek™ RTX™-1 Capillary Columns, Under 30m, Particle Size Greater Than 1.50μm: 10m Length DFS Item
General-purpose columns for solvent impurities
Specifications
Coating Fused Silica
Packing Material Rtx™-1
Technique Crossbond™
For Use With Solvent impurities, PCB congeners, simulated distillation, drugs of abuse, gases, natural gas odorants, Sulfur compounds, essential oils, Hydrocarbons, semivolatiles, pesticides, Oxygenates analysis
Material Stainless Steel
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Particle Size Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Particle Size Price Quantity  
06708466 View Documents Restek™
RK10182107
0.53mm 3μm 10m 3μm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Nonpolar phase; Crossbond™ dimethyl polysiloxane,
  • PCB congeners (e.g. Aroclor mixes), simulated distillation, arson accelerants, gases, natural gas odorants, sulfur compounds, essential oils, hydrocarbons, semivolatiles, pesticides, oxygenates
  • Temperature range: -60°C to 350°C (-76°F to 662°F)
  • Equivalent to USP G1, G2, G38 phases
Specifications
Specifications
Fused Silica
Crossbond™
Stainless Steel
Non polar
G1, G2, G38
Rtx™-1
Solvent impurities, PCB congeners, simulated distillation, drugs of abuse, gases, natural gas odorants, Sulfur compounds, essential oils, Hydrocarbons, semivolatiles, pesticides, Oxygenates analysis
350°C
100% Dimethyl Polysiloxane