Restek™ Rtx™-1 Capillary Columns DFS Item
$1,527.39 - $1,591.07
Specifications
Coating Fused Silica
Film Thickness 0.1 µm
Packing Material Rtx™-1
For Use With (Application) Arson Accelerants,Essential Oils,Gases Analysis,Hydrocarbons,Natural Gas Odorants Analysis,Oxygenates Analysis,PCB Congeners (e.g. Aroclor Mixes),Pesticide Analysis,Semivolatiles Analysis,Simulated Distillation,Solvent Impurities Analysis,Sulfur Compounds Analysis
Max. Temperature 350°C
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Includes Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Includes Price Quantity  
06708220 View Documents Restek™
RK10105124
0.25 mm 15 m 5m Integra-Guard Column N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06708230 View Documents Restek™
RK10108112
0.25 mm 20 m N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06708231 View Documents Restek™
RK10108124
0.25 mm 30 m 5m Integra-Guard Column N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06708234 View Documents Restek™
RK10109106
0.32 mm 25 m Each for $1,527.39
06708235 View Documents Restek™
RK10109125
0.32 mm 30 m 5m Integra-Guard Column Each for $1,591.07
Specifications
Specifications
Fused Silica
Rtx™-1
350°C
100% Dimethyl Polysiloxane
0.1 µm
Arson Accelerants,Essential Oils,Gases Analysis,Hydrocarbons,Natural Gas Odorants Analysis,Oxygenates Analysis,PCB Congeners (e.g. Aroclor Mixes),Pesticide Analysis,Semivolatiles Analysis,Simulated Distillation,Solvent Impurities Analysis,Sulfur Compounds Analysis
Non polar
G1, G2, G38