Restek™ Rtx™-1 Capillary Columns DFS Item
Manufacturer:  Restek™ RK10109125
 View more versions of this product
Catalog No. 06708235
Specifications
Specifications
Solvent Impurities, PCB Congeners (e.g. Aroclor Mixes), Simulated Distillation, Arson Accelerants, Gases, Natural Gas Odorants, Sulfur Compounds, Essential Oils, Hydrocarbons, Semivolatiles, Pesticides, Oxygenates
G1, G2, G38
0.1μm
Crossbond™
Rtx™-1
Fused Silica
5m Integra-Guard Column
350°C
-60 to 330/350°C
30m
100% Dimethyl Polysiloxane
Non Polar
0.32mm
Documents