Restek™ Molecular Sieve Packed Columns: SilcoSmooth Tubing DFS Item
Easily separate permanent gases at above-ambient temperatures
$405.33 - $889.28
Specifications
Diameter (Metric) Inner 2 mm
Diameter (Metric) Outer 3.17 mm
View More Specs
Products 40
Catalog Number Mfr. No. Length (Metric) Mesh Size Description Packing Material Price Quantity  
Catalog Number Mfr. No. Length (Metric) Mesh Size Description Packing Material Price Quantity  
06714533
View Documents
Restek™
RK80428800
2 m 60/80 General Configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714534
View Documents
Restek™
RK80428810
2 m 60/80 Configured for Hewlett Packard Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714535
View Documents
Restek™
RK80428820
2 m 60/80 Configured for Varian/Vista Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714536
View Documents
Restek™
RK80428830
2 m 60/80 Configured for PE/Sigma Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714537
View Documents
Restek™
RK80428840
2 m 60/80 Configured for PE Auto system Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714538
View Documents
Restek™
RK80429800
2 m 80/100 General Configuration Mole Sieve 5Å Each for $684.78
06714539
View Documents
Restek™
RK80429800901
2 m 80/100 General configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714540
View Documents
Restek™
RK80429810
2 m 80/100 Configured for Hewlett Packard Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714541
View Documents
Restek™
RK80429820
2 m 80/100 Configured for Varian/Vista Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714543
View Documents
Restek™
RK80429840
2 m 80/100 Configured for PE Auto system Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714544
View Documents
Restek™
RK80429865901
2 m 80/100 Carlo Erba configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714545
View Documents
Restek™
RK80429890
2 m 80/100 Gow Mac 590 configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714546
View Documents
Restek™
RK80429895
2 m 80/100 SRI 8610C configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714547
View Documents
Restek™
RK80429895R
2 m 80/100 SRI 8610C dual GC right side configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714548
View Documents
Restek™
RK80430800
3.05 m 80/100 General Configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714549
View Documents
Restek™
RK80430810
3.05 m 80/100 Configured for Hewlett Packard Mole Sieve 5Å Each for $889.28
06714550
View Documents
Restek™
RK80430810901
3.05 m 80/100 Agilent 5880, 5890, 5987, 6890, 7890 configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714551
View Documents
Restek™
RK80430820
3.05 m 80/100 Configured for Varian/Vista Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714552
View Documents
Restek™
RK80430830
3.05 m 80/100 Configured for PE/Sigma Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714553
View Documents
Restek™
RK80430840
3.05 m 80/100 Configured for PE Auto system Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714554
View Documents
Restek™
RK80430840901
3.05 m 80/100 PerkinElmer Auto System 8300, 8400, 8700, Clarus 500 (C500) configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714555
View Documents
Restek™
RK80430895
3.05 m 80/100 SRI 8610C configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714581
View Documents
Restek™
RK80439800
2 m 80/100 General Configuration Mole Sieve 13X Each for $664.59
06714582
View Documents
Restek™
RK80439810
2 m 80/100 Configured for Agilent Mole Sieve 13X Each for $664.59
06714583
View Documents
Restek™
RK80440800
1 m 80/100 General Configuration Mole Sieve 5Å Each for $480.30
06714584
View Documents
Restek™
RK80440810
1 m 80/100 Configured for Agilent Mole Sieve 5Å Each for $480.30
06714585
View Documents
Restek™
RK80440810901
1 m 80/100 Agilent 5880, 5890, 5987, 6890, 7890 configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714656
View Documents
Restek™
RK80473800
1.8 m 60/80 General Configuration Mole Sieve 5Å Each for $537.22
06714658
View Documents
Restek™
RK80473810901
1.82 m 60/80 Agilent 5880, 5890, 5987, 6890, 7890 configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714657
View Documents
Restek™
RK80473810
1.8 m 60/80 Configured for Agilent Mole Sieve 5Å Each for $537.22
06714661
View Documents
Restek™
RK80474810901
1.82 m 80/100 Agilent 5880, 5890, 5987, 6890, 7890 configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714663
View Documents
Restek™
RK80474840901
1.82 m 80/100 PerkinElmer Auto System 8300, 8400, 8700, Clarus 500 (C500) configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714664
View Documents
Restek™
RK80474845
1.82 m 80/100 ABB 3100, AAI (4 in. coil) configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714666
View Documents
Restek™
RK80475800901
1.82 m 60/80 General configuration Mole Sieve 13X N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714680
View Documents
Restek™
RK80480800
2 m 60/80 General configuration Mole Sieve 13X Each for $662.74
06714706
View Documents
Restek™
RK88014800
3.04 m 80/100 General configuration Mole Sieve 5Å Each for $739.33
06714707
View Documents
Restek™
RK88014810
3.04 m 80/100 Configured for Hewlett Packard Mole Sieve 5Å Each for $739.33
06714708
View Documents
Restek™
RK88014820
3.04 m 80/100 Configured for Varian Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06714711
View Documents
Restek™
RK88015810
0.9 m 80/100 Configured for Agilent Mole Sieve 5Å Each for $405.33
06714710
View Documents
Restek™
RK88015800
0.9 m 80/100 General configuration Mole Sieve 5Å N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Excellent batch-to-batch reproducibility
  • Preactivated and ready to use
Order Info
Each column comes with metal end-fittings to prevent water or carbon dioxide from adsorbing into the packing during shipment.
Specifications
Specifications
2 mm
3.17 mm