VICI™ Restek™ Gas-Specific Purifiers DFS Item
Ideal for removing moisture, hydrocarbons and oxygen from gas chromatography gas supplies
Manufacturer:  VICI™ MSP1001
 View more versions of this product
Catalog No. 05-730-1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Used in-line with the GC carrier gas or detector gas supply to produce gases that are at least a factor of ten higher in purity than a 99.9999% “Chromatography Grade” cylinder of gas (when the purifier is supplied by a 99.995% “High Purity” cylinder of gas).
  • Available with 1/8 in. or 1/4 in. compression fittings
  • All models feature proprietary adsorptive materials to help capture and retain specific contaminants for the operating life of the purifier
  • These materials adsorb a larger number and variety of contaminants than commonly used adsorptive materials
Specifications
Specifications
Helium/Inert
Gas Purifier
0.12 in. Fitting
22.5 in.
3.8 cm
500 mL/min.
1000 psi
57.15 cm
1.5 in.
Ideal for removing moisture, Hydrocarbons and Oxygen from gas chromatography gas supplies