Restek™ 20 mm Aluminum Seals with Septa, Assembled DFS Item
Preassembled seals save time and increase productivity
Supplier:  Restek™ RK21763
 View more versions of this product
Catalog No. 06718652
Description
Description
  • Color: Silver
  • Material: PTFE/Gray Butyl Rubber Septa
Specifications
Specifications
Silver
PTFE, Silicone
Crimp Cap
Aluminum
100
20 mm