Please login to your online account to display your discounted pricing

Resorcinol TS, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Resorcinol
Name Note Test Solution
CAS 7647-01-0
CAS Min % 97.0
CAS Max % 100.0
CAS 108-46-3
CAS Min % 0.82
CAS Max % 0.86
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6619-16

 
ricca chemical company
RI661916
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

6619-4

 
ricca chemical company
RI66194
Glass amber 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless to slight greenish yellow
Physical Form Liquid
Quantity 500mL
Chemical Name or Material Resorcinol
Name Note Test Solution
CAS Min % 97.0
CAS 7647-01-0
CAS Max % 100.0
CAS 108-46-3
Packaging Glass amber
CAS Min % 0.82
CAS Max % 0.86