Please login to your online account to display your discounted pricing

Alfa Aesar™ Resorcinol-O,O'-diacetic acid, 97+%

$64.03 - $64.03

Chemical Identifiers

Chemical Name or Material Resorcinol-O,O'-diacetic acid
CAS 102-39-6
Molecular Formula C10H10O6
Formula Weight 226.19
MDL Number MFCD00016696
Beilstein 2135777
Synonym 1,3-Bis(carboxymethoxy)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0550503

 
alfa aesar™
L0550503
1g Each for $64.03
Description & Specifications

Specifications

Formula Weight 226.19
Chemical Name or Material Resorcinol-O,O'-diacetic acid
Molecular Formula C10H10O6
MDL Number MFCD00016696
CAS 102-39-6
Synonym 1,3-Bis(carboxymethoxy)benzene
Assay ≥97%
Beilstein 2135777
Melting Point 192° to 196°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.