Please login to your online account to display your discounted pricing

Alfa Aesar™ Resorcinol monobenzoate, 94%

$32.26 - $105.03

Chemical Identifiers

Chemical Name or Material Resorcinol monobenzoate
CAS 136-36-7
Molecular Formula C13H10O3
Formula Weight 214.22
MDL Number MFCD00020118
Synonym 3-Benzoyloxyphenol; 3-Hydroxyphenyl benzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1915036

  SDS
alfa aesar™
A1915036
500g Each for $105.03

AAA1915022

  SDS
alfa aesar™
A1915022
100g Each for $32.26
Description & Specifications

Specifications

Formula Weight 214.22
Chemical Name or Material Resorcinol monobenzoate
TSCA TSCA
Assay 94%
Molecular Formula C13H10O3
MDL Number MFCD00020118
CAS 136-36-7
Synonym 3-Benzoyloxyphenol; 3-Hydroxyphenyl benzoate
Melting Point 130° to 133°C
Health Hazard 1 Warning
Quantity 500g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VH2800000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.