Resorcinol, Crystalline Powder, USP, 99-100.5%, Spectrum™ Chemical
R1005, 108-46-3, C6H6O2
Supplier:  Spectrum Chemical Mfg Cor R1005125GM
 View more versions of this product
Catalog No. 18606066
Description
Description
Spectrum™ Resorcinol, Crystalline Powder, USP is used as an antiseptic and disinfectant and aid in treating skin diseases such as eczema. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
108-46-3
0.0005
109-111°C
C6H6O2
GHMLBKRAJCXXBS-UHFFFAOYSA-N
benzene-1,3-diol
99 to 100.5%
1
0.01
Amber Glass Bottle
125 g
OC1=CC(O)=CC=C1
110.11
USP
Chemical Identifiers
108-46-3
110.11
benzene-1,3-diol
C6H6O2
GHMLBKRAJCXXBS-UHFFFAOYSA-N
OC1=CC(O)=CC=C1