Please login to your online account to display your discounted pricing

Resorcinol 98%, ACROS Organics™


Quantity 1kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC132290010

 View more versions of this product

Catalog No. AC132290010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Melting Point 108° to 112°C
Formula Weight 110.11
Physical Form Liquid
Quantity 1kg
Color Clear colorless to light yellow
RTECS Number VG9625000
EINECS Number 203-585-2
TSCA TSCA
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Sulfated Ash 0.1% max.
Packaging Plastic bottle
Synonym 1, 3-Dihydroxybenzene
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Name Note 98%
MDL Number MFCD00002269
Assay Percent Range 98%
Molecular Formula C6H6O2
CAS 108-46-3
Chemical Name or Material Resorcinol

Specifications

Chemical Name or Material Resorcinol
Name Note 98%
CAS 108-46-3
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H6O2
Formula Weight 110.11
MDL Number MFCD00002269
Synonym 1, 3-Dihydroxybenzene
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

SDS & Safety Documents