Please login to your online account to display your discounted pricing

Resiquimod, Tocris™

Toll-like receptor 7 (TLR7) agonist

$116.01 - $447.38

Chemical Identifiers

Chemical Name or Material Resiquimod
Assay Percent Range >98%
Molecular Formula C17H22N4O2
Formula Weight 314.38
Synonym 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4, 5-c]quinolin-1-yl]-2-methylpropan-2-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

453610

 
r & d systems
453610
10mg Each for $116.01

453650

 
r & d systems
453650
50mg Each for $447.38
Description & Specifications

Specifications

Formula Weight 314.38
Quantity 10mg
Chemical Name or Material Resiquimod
Assay Percent Range >98%
Molecular Formula C17H22N4O2
CAS 144875-48-9
Synonym 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4, 5-c]quinolin-1-yl]-2-methylpropan-2-ol
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C