Please login to your online account to display your discounted pricing

Resiniferatoxin, 99%, high purity, ACROS Organics™


Quantity 5mg

Manufacturer: thermo fisher scientific  AC328470050

 View more versions of this product

Catalog No. AC328470050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Solid
Formula Weight 628.7
Quantity 5mg
Color White to off-white
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material Resiniferatoxin
Molecular Formula C37H40O9
Name Note High Purity
MDL Number MFCD00135927
Assay Percent Range 99%
CAS 57444-62-9

Specifications

Chemical Name or Material Resiniferatoxin
Name Note High Purity
CAS 57444-62-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C37H40O9
Formula Weight 628.7
MDL Number MFCD00135927
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents