missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Resiniferatoxin, 99%, high purity, ACROS Organics™

Chemical Identifiers

CAS 57444-62-9
Molecular Formula C37H40O9
Molecular Weight (g/mol) 628.718
MDL Number MFCD00135927
InChI Key DSDNAKHZNJAGHN-BWVJFJMLSA-N
Synonym resiniferatoxin
PubChem CID 57090162
SMILES CC1CC2(C3C4C1(C5C=C(C(=O)C5(CC(=C4)COC(=O)CC6=CC(=C(C=C6)O)OC)O)C)OC(O3)(O2)CC7=CC=CC=C7)C(=C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC328470050 SDS View Documents Acros Organics
AC328470050
5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC328470010 SDS View Documents Acros Organics
328470010
1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

57444-62-9
628.718
DSDNAKHZNJAGHN-BWVJFJMLSA-N
57090162
C37H40O9
MFCD00135927
resiniferatoxin
CC1CC2(C3C4C1(C5C=C(C(=O)C5(CC(=C4)COC(=O)CC6=CC(=C(C=C6)O)OC)O)C)OC(O3)(O2)CC7=CC=CC=C7)C(=C)C

Specifications

White
57444-62-9
C37H40O9
resiniferatoxin
CC1CC2(C3C4C1(C5C=C(C(=O)C5(CC(=C4)COC(=O)CC6=CC(=C(C=C6)O)OC)O)C)OC(O3)(O2)CC7=CC=CC=C7)C(=C)C
57090162
Solid
Resiniferatoxin, High Purity
5mg
99%
MFCD00135927
DSDNAKHZNJAGHN-BWVJFJMLSA-N
628.718
628.7
99%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.